siRNA Details
| Gene Symbol: Grb10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109093.1 | NP_001102563.1 | 3 | 208 | ||
| XM_006251508.2 | XP_006251570.1 | 2 | 161 | |||
| XM_008770347.1 | XP_008768569.1 | 3 | 326 | |||
| XM_008770348.1 | XP_008768570.1 | 3 | 326 | |||

Grb10
Genomic Map