siRNA Details
| Gene Symbol: RGD1562229 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109095.1 | NP_001102565.1 | 2 | 111 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1562229| Gene Symbol: RGD1562229 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109095.1 | NP_001102565.1 | 2 | 111 | ||