siRNA Details
| Gene Symbol: Psme4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025140.2 | NP_001020311.2 | 31, 32 | 3854 | ||
| XM_008770467.1 | XP_008768689.1 | 32, 33 | 4170 | |||
| GenBank mRNA | CA339363.1 | - | 3, 4 | 335 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Psme4| Gene Symbol: Psme4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025140.2 | NP_001020311.2 | 31, 32 | 3854 | ||
| XM_008770467.1 | XP_008768689.1 | 32, 33 | 4170 | |||
| GenBank mRNA | CA339363.1 | - | 3, 4 | 335 | ||