siRNA Details
| Gene Symbol: Gpr75 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109096.1 | NP_001102566.1 | 1 | 677 | ||
| XM_006251616.2 | XP_006251678.1 | 1 | 782 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpr75| Gene Symbol: Gpr75 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109096.1 | NP_001102566.1 | 1 | 677 | ||
| XM_006251616.2 | XP_006251678.1 | 1 | 782 | |||
Set Membership: |
|