siRNA Details
| Gene Symbol: LOC498460 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047931.1 | NP_001041396.1 | 3 | 275 | ||
| GenBank mRNA | AY383710.1 | - | 3 | 275 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC498460| Gene Symbol: LOC498460 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047931.1 | NP_001041396.1 | 3 | 275 | ||
| GenBank mRNA | AY383710.1 | - | 3 | 275 | ||