siRNA Details
| Gene Symbol: Ptgdr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135164.1 | NP_001128636.1 | 2 | 990 | ||
| GenBank mRNA | AF120101.1 | - | 2 | 990 | ||
| FQ222068.1 | - | 1092 | ||||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ptgdr| Gene Symbol: Ptgdr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135164.1 | NP_001128636.1 | 2 | 990 | ||
| GenBank mRNA | AF120101.1 | - | 2 | 990 | ||
| FQ222068.1 | - | 1092 | ||||
Set Membership: |
|