siRNA Details
| Gene Symbol: Gnpnat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006251782.2 | XP_006251844.1 | 1, 2 | 93 | ||
| XM_006251784.2 | XP_006251846.1 | 1, 2 | 89 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gnpnat1| Gene Symbol: Gnpnat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006251782.2 | XP_006251844.1 | 1, 2 | 93 | ||
| XM_006251784.2 | XP_006251846.1 | 1, 2 | 89 | |||