siRNA Details
| Gene Symbol: RGD1565212 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109103.1 | NP_001102573.1 | 2 | 255 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_026359, AK005778, AK016302, BC048583.

RGD1565212
Genomic Map