siRNA Details
Gene Symbol: RGD1565212 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109103.1 | NP_001102573.1 | 4 | 766 |

More Information
Other Species Targeted


Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RGD1565212 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109103.1 | NP_001102573.1 | 4 | 766 |