siRNA Details
| Gene Symbol: Serp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109104.1 | NP_001102574.1 | 4 | 630 | ||
| XM_008770884.1 | XP_008769106.1 | 4 | 362 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Serp2| Gene Symbol: Serp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109104.1 | NP_001102574.1 | 4 | 630 | ||
| XM_008770884.1 | XP_008769106.1 | 4 | 362 | |||