siRNA Details
Gene Symbol: Spert | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017506.1 | NP_001017506.1 | 2 | 170 | ||
XM_006252329.2 | XP_006252391.1 | 2 | 174 | |||
XM_008770893.1 | XP_008769115.1 | 4 | 2610 | |||
XM_008770894.1 | XP_008769116.1 | 3 | 218 | |||
XM_008770895.1 | XP_008769117.1 | 3 | 211 | |||
GenBank mRNA | BC079300.1 | - | 2 | 170 | 7134526 |
