siRNA Details
| Gene Symbol: Ldb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277165.1 | NP_001264094.1 | 10 | 1488 | ||
| NM_001277166.1 | NP_001264095.1 | 11 | 1347 | |||
| XM_006252637.2 | XP_006252699.1 | 10 | 1544 | |||
| XM_006252640.2 | XP_006252702.1 | 11 | 1403 | |||
| XM_006252641.2 | XP_006252703.1 | 9 | 1358 | |||
| XM_006252644.2 | XP_006252706.1 | 10 | 1217 | |||

Ldb3
Genomic Map