siRNA Details
| Gene Symbol: RGD1559804 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113787.1 | NP_001107259.1 | 3 | 614 | ||
| GenBank mRNA | BC158776.1 | - | 3 | 614 | 7133601 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1559804| Gene Symbol: RGD1559804 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113787.1 | NP_001107259.1 | 3 | 614 | ||
| GenBank mRNA | BC158776.1 | - | 3 | 614 | 7133601 | |