siRNA Details
| Gene Symbol: Lrit2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191938.1 | NP_001178867.1 | 2 | 723 | ||
| XM_006252787.2 | XP_006252849.1 | 2 | 2006 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrit2| Gene Symbol: Lrit2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191938.1 | NP_001178867.1 | 2 | 723 | ||
| XM_006252787.2 | XP_006252849.1 | 2 | 2006 | |||