siRNA Details
| Gene Symbol: STK17B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004226.3 | NP_004217.1 | 7 | 1033 | ||
| XM_011512168.2 | XP_011510470.1 | 7 | 998 | |||
| XM_011512169.1 | XP_011510471.1 | 7 | 1031 | |||
| XM_011512170.1 | XP_011510472.1 | 8 | 1014 | |||
| XM_011512171.2 | XP_011510473.1 | 6 | 526 | |||
| GenBank mRNA | AB011421.1 | - | 7 | 970 | ||
| AK312865.1 | - | 7 | 1031 | |||
| BC016040.1 | - | 7 | 972 | 4575012 | ||
| CR542210.1 | - | 6 | 709 | |||
| CR542225.1 | - | 6 | 709 | |||

STK17B
Genomic Map