siRNA Details
| Gene Symbol: Fam129c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100908.1 | NP_001094378.1 | 3 | 283 | ||
| GenBank mRNA | BC111077.1 | - | 3 | 283 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam129c| Gene Symbol: Fam129c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100908.1 | NP_001094378.1 | 3 | 283 | ||
| GenBank mRNA | BC111077.1 | - | 3 | 283 | ||