siRNA Details
| Gene Symbol: Kxd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025143.1 | NP_001020314.1 | 3 | 248 | ||
| XM_006252922.2 | XP_006252984.1 | 3 | 326 | |||
| XM_006252923.2 | XP_006252985.1 | 3 | 653 | |||
| XM_006252924.1 | XP_006252986.1 | 3 | 593 | |||
| XM_006252925.1 | XP_006252987.1 | 190 | ||||
| GenBank mRNA | BC088216.1 | - | 3 | 248 | 7383995 | |

Kxd1
Genomic Map