siRNA Details
| Gene Symbol: Tceb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270561.1 | NP_001257490.1 | 4 | 728 | ||
| NM_001270562.1 | NP_001257491.1 | 4 | 626 | |||
| NM_001270563.1 | NP_001257492.1 | 4 | 586 | |||
| NM_022593.4 | NP_072115.1 | 4 | 622 | |||
| XM_006237724.2 | XP_006237786.1 | 4 | 623 | |||
| GenBank mRNA | BC060566.1 | - | 4 | 597 | 6918038 | |
| CK355540.1 | - | 4 | 622 | |||
| FM093546.1 | - | 4 | 588 | |||
| FM101371.1 | - | 4 | 576 | |||
| FM110992.1 | - | 4 | 592 | |||
| FQ213660.1 | - | 4 | 636 | |||
| FQ214805.1 | - | 4 | 728 | |||

Tceb1
Genomic Map