siRNA Details
| Gene Symbol: Rbpms | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271244.1 | NP_001258173.1 | 3 | 451 | ||
| XM_006253238.2 | XP_006253300.1 | 3 | 469 | |||
| XM_006253240.2 | XP_006253302.1 | 3 | 466 | |||
| XM_006253241.2 | XP_006253303.1 | 3 | 469 | |||
| XM_008771303.1 | XP_008769525.1 | 3 | 469 | |||

Rbpms
Genomic Map