siRNA Details
| Gene Symbol: Rab11fip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191555.1 | NP_001178484.1 | 6 | 3421 | ||
| NM_001197241.1 | NP_001184170.1 | 5 | 1798 | |||
| XM_008771331.1 | XP_008769553.1 | 6 | 3678 | |||
| GenBank mRNA | BC107469.1 | - | 5 | 1798 | 7458514 | |
| FQ221846.1 | - | 5 | 1807 | |||

Rab11fip1
Genomic Map