siRNA Details
| Gene Symbol: Tdrp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100791.1 | NP_001094261.1 | 3, 4 | 330 | ||
| XM_006253397.2 | XP_006253459.2 | 3, 4 | 388 | |||
| XM_006253398.2 | XP_006253460.2 | 3, 4 | 364 | |||
| XM_006253399.1 | XP_006253461.1 | 3, 4 | 404 | |||
| GenBank mRNA | BC100251.1 | - | 3, 4 | 313 | ||
| BC168702.1 | - | 3, 4 | 330 | 7455266 | ||

Tdrp
Genomic Map