siRNA Details
| Gene Symbol: Fam155a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751632.3 | XP_003751680.1 | 4 | 1343 | ||
| XM_573950.6 | XP_573950.4 | 3 | 1297 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam155a| Gene Symbol: Fam155a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751632.3 | XP_003751680.1 | 4 | 1343 | ||
| XM_573950.6 | XP_573950.4 | 3 | 1297 | |||