siRNA Details
| Gene Symbol: Spry3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008769065.1 | XP_008767287.1 | 2 | 2634 | ||
| XM_008769066.1 | XP_008767288.1 | 3 | 1555 | |||
| XM_008769067.1 | XP_008767289.1 | 3 | 1507 | |||
| XM_008769068.1 | XP_008767290.1 | 2 | 1223 | |||
| XM_008769069.1 | XP_008767291.1 | 2 | 1353 | |||
| GenBank mRNA | BC168971.1 | - | 2 | 1253 | 7316797 | |
| Gene Symbol: RGD1564382 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008758720.1 | XP_008756942.1 | 1 | 743 | ||

Spry3
Genomic Map