siRNA Details
| Gene Symbol: Aaed1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006253495.2 | XP_006253557.1 | 6 | 643 | ||
| XM_006253497.2 | XP_006253559.1 | 5 | 515 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aaed1| Gene Symbol: Aaed1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006253495.2 | XP_006253557.1 | 6 | 643 | ||
| XM_006253497.2 | XP_006253559.1 | 5 | 515 | |||