siRNA Details
| Gene Symbol: MGC114246 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017509.1 | NP_001017509.1 | 7 | 944 | ||
| GenBank mRNA | BC091563.1 | - | 7 | 944 | 7461240 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

MGC114246| Gene Symbol: MGC114246 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017509.1 | NP_001017509.1 | 7 | 944 | ||
| GenBank mRNA | BC091563.1 | - | 7 | 944 | 7461240 | |