siRNA Details
| Gene Symbol: Fam193b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170408.1 | NP_001163879.1 | 3, 4 | 422 | ||
| XM_006253646.2 | XP_006253708.1 | 2, 3 | 521 | |||
| XM_006253650.2 | XP_006253712.1 | 2, 3 | 1155 | |||
| XM_008771503.1 | XP_008769725.1 | 2, 3 | 2940 | |||
| GenBank mRNA | CB719022.1 | - | 3, 4 | 422 | ||

Fam193b
Genomic Map