siRNA Details
| Gene Symbol: Fam193b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170408.1 | NP_001163879.1 | 8 | 2023 | ||
| XM_006253646.2 | XP_006253708.1 | 7 | 2122 | |||
| XM_006253647.2 | XP_006253709.1 | 7 | 2862 | |||
| XM_006253650.2 | XP_006253712.1 | 7 | 2756 | |||
| XM_006253651.2 | XP_006253713.1 | 8 | 2923 | |||
| XM_008771503.1 | XP_008769725.1 | 7 | 4541 | |||
| GenBank mRNA | BC166819.1 | - | 2 | 816 | 8367512 | |

Fam193b
Genomic Map