siRNA Details
| Gene Symbol: Prr7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109116.1 | NP_001102586.1 | 2 | 929 | ||
| XM_006253652.2 | XP_006253714.1 | 2 | 1173 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prr7| Gene Symbol: Prr7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109116.1 | NP_001102586.1 | 2 | 929 | ||
| XM_006253652.2 | XP_006253714.1 | 2 | 1173 | |||