siRNA Details
| Gene Symbol: LOC102554838 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006253772.2 | XP_006253834.1 | 2 | 450 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102554838| Gene Symbol: LOC102554838 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006253772.2 | XP_006253834.1 | 2 | 450 | ||