siRNA Details
| Gene Symbol: LOC102554838 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006253772.2 | XP_006253834.1 | 4 | 713 | ||
| Gene Symbol: Stmnd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008771561.1 | XP_008769783.1 | 207 | |||

LOC102554838
Genomic Map