siRNA Details
| Gene Symbol: Elovl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109118.1 | NP_001102588.1 | 7, 8 | 884 | ||
| XM_006253779.2 | XP_006253841.1 | 7, 8 | 830 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Elovl2| Gene Symbol: Elovl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109118.1 | NP_001102588.1 | 7, 8 | 884 | ||
| XM_006253779.2 | XP_006253841.1 | 7, 8 | 830 | |||