siRNA Details
| Gene Symbol: Tpi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022922.2 | NP_075211.2 | 7 | 1212 | ||
| GenBank mRNA | BC126087.1 | - | 7 | 1212 | 8368624 | |
| FQ215213.1 | - | 6 | 1127 | |||
| FQ216226.1 | - | 6 | 1150 | |||
| FQ216307.1 | - | 7 | 1111 | |||
| FQ223237.1 | - | 7 | 1226 | |||
| FQ224920.1 | - | 7 | 1227 | |||
| Gene Symbol: LOC100911515 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750654.3 | XP_003750702.2 | 7 | 1368 | ||

Tpi1
Genomic Map