siRNA Details
| Gene Symbol: Fam217a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271083.1 | NP_001258012.1 | 2 | 153 | ||
| GenBank mRNA | BC083911.1 | - | 548 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam217a| Gene Symbol: Fam217a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271083.1 | NP_001258012.1 | 2 | 153 | ||
| GenBank mRNA | BC083911.1 | - | 548 | |||