siRNA Details
| Gene Symbol: RGD1561310 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008771609.1 | XP_008769831.1 | 1 | 63 | ||
| XM_574022.5 | XP_574022.3 | 1 | 63 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1561310| Gene Symbol: RGD1561310 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008771609.1 | XP_008769831.1 | 1 | 63 | ||
| XM_574022.5 | XP_574022.3 | 1 | 63 | |||