siRNA Details
| Gene Symbol: BCL7C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004765.3 | NP_004756.2 | 6 | 902 | ||
| XM_017023885.1 | XP_016879374.1 | 6 | 1153 | |||
| GenBank mRNA | AJ223980.1 | - | 6 | 544 | ||
| BC019071.1 | - | 6 | 678 | 4909589 | ||

BCL7C
Genomic Map