siRNA Details
| Gene Symbol: Fam107b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025034.1 | NP_001020205.1 | 2 | 299 | ||
| XM_008771879.1 | XP_008770101.1 | 2 | 698 | |||
| XM_008771880.1 | XP_008770102.1 | 2 | 321 | |||
| XM_008771881.1 | XP_008770103.1 | 2 | 747 | |||
| XM_008771882.1 | XP_008770104.1 | 2 | 334 | |||
| XM_008771883.1 | XP_008770105.1 | 2 | 951 | |||
| XM_008771884.1 | XP_008770106.1 | 2 | 343 | |||
| GenBank mRNA | BC085116.1 | - | 2 | 299 | 7107468 | |

Fam107b
Genomic Map