siRNA Details
| Gene Symbol: Thnsl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025035.1 | NP_001020206.1 | 4 | 1937 | ||
| GenBank mRNA | BC091213.1 | - | 4 | 1937 | 7382276 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Thnsl1| Gene Symbol: Thnsl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025035.1 | NP_001020206.1 | 4 | 1937 | ||
| GenBank mRNA | BC091213.1 | - | 4 | 1937 | 7382276 | |