siRNA Details
| Gene Symbol: Myo3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_574090.5 | XP_574090.4 | 24 | 2977 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Myo3a| Gene Symbol: Myo3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_574090.5 | XP_574090.4 | 24 | 2977 | ||