siRNA Details
| Gene Symbol: Greb1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008772006.1 | XP_008770228.1 | 13, 14 | 3217 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Greb1l| Gene Symbol: Greb1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008772006.1 | XP_008770228.1 | 13, 14 | 3217 | ||