siRNA Details
| Gene Symbol: MGC116121 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024905.1 | NP_001020076.1 | 3 | 293 | ||
| XM_008771953.1 | XP_008770175.1 | 3 | 372 | |||
| GenBank mRNA | BC097984.1 | - | 3 | 293 | 7444421 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

MGC116121| Gene Symbol: MGC116121 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024905.1 | NP_001020076.1 | 3 | 293 | ||
| XM_008771953.1 | XP_008770175.1 | 3 | 372 | |||
| GenBank mRNA | BC097984.1 | - | 3 | 293 | 7444421 | |