siRNA Details
| Gene Symbol: RGD1562608 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751756.3 | XP_003751804.2 | 1, 2 | 315 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1562608| Gene Symbol: RGD1562608 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751756.3 | XP_003751804.2 | 1, 2 | 315 | ||