siRNA Details
Gene Symbol: Rps27l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772051.1 | XP_008770273.1 | 1 | 157 | ||
XM_574121.5 | XP_574121.1 | 1 | 155 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rps27l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772051.1 | XP_008770273.1 | 1 | 157 | ||
XM_574121.5 | XP_574121.1 | 1 | 155 |