siRNA Details
| Gene Symbol: Pcdhga2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037139.1 | NP_001032216.1 | 1 | 1099 | ||
| GenBank mRNA | AY574012.1 | - | 1 | 1099 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pcdhga2| Gene Symbol: Pcdhga2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037139.1 | NP_001032216.1 | 1 | 1099 | ||
| GenBank mRNA | AY574012.1 | - | 1 | 1099 | ||