siRNA Details
| Gene Symbol: Scgb3a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024283.1 | NP_001019454.1 | 2 | 172 | ||
| GenBank mRNA | AF439547.1 | - | 2 | 172 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Scgb3a2| Gene Symbol: Scgb3a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024283.1 | NP_001019454.1 | 2 | 172 | ||
| GenBank mRNA | AF439547.1 | - | 2 | 172 | ||