siRNA Details
| Gene Symbol: LOC103694226 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008772196.1 | XP_008770418.1 | 1 | 1175 | ||
| Gene Symbol: LOC102555392 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006254757.2 | XP_006254819.1 | 1175 | |||
| Gene Symbol: MGC108823 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012353.1 | NP_001012353.1 | 2 | 1293 | ||
| GenBank mRNA | BC089836.1 | - | 2 | 1293 | 7326302 | |

LOC103694226
Genomic Map