siRNA Details
| Gene Symbol: RGD1560212 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077676.1 | NP_001071144.1 | 4 | 944 | ||
| XM_006254957.2 | XP_006255019.1 | 3 | 899 | |||
| XM_008772164.1 | XP_008770386.1 | 4 | 920 | |||
| GenBank mRNA | BC127540.1 | - | 4 | 944 | 7301991 | |
| FQ234858.1 | - | 3 | 909 | |||
| Gene Symbol: LOC100909833 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003752266.2 | XP_003752314.1 | 557 | |||

RGD1560212
Genomic Map