siRNA Details
Gene Symbol: LOC498933 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025764.1 | NP_001020935.1 | 5 | 585 | ||
GenBank mRNA | BC098038.1 | - | 5 | 585 | 7456623 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC498933 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025764.1 | NP_001020935.1 | 5 | 585 | ||
GenBank mRNA | BC098038.1 | - | 5 | 585 | 7456623 |