siRNA Details
| Gene Symbol: Fbxl8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109128.1 | NP_001102598.1 | 3 | 588 | ||
| XM_006255520.2 | XP_006255582.1 | 3 | 358 | |||
| XM_006255521.2 | XP_006255583.1 | 2 | 223 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fbxl8| Gene Symbol: Fbxl8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109128.1 | NP_001102598.1 | 3 | 588 | ||
| XM_006255520.2 | XP_006255582.1 | 3 | 358 | |||
| XM_006255521.2 | XP_006255583.1 | 2 | 223 | |||