siRNA Details
| Gene Symbol: RGD1561415 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109129.1 | NP_001102599.1 | 3 | 529 | ||
| XM_006255523.2 | XP_006255585.1 | 3 | 552 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1561415| Gene Symbol: RGD1561415 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109129.1 | NP_001102599.1 | 3 | 529 | ||
| XM_006255523.2 | XP_006255585.1 | 3 | 552 | |||