siRNA Details
| Gene Symbol: Il34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025766.1 | NP_001020937.1 | 3 | 575 | ||
| XM_006255599.2 | XP_006255661.1 | 4 | 938 | |||
| XM_006255600.2 | XP_006255662.1 | 4 | 938 | |||
| XM_008772527.1 | XP_008770749.1 | 4 | 643 | |||
| GenBank mRNA | BC076391.1 | - | 4 | 701 | ||
| BC098808.1 | - | 3 | 575 | 7376793 | ||

Il34
Genomic Map